missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Hayward PP Needle Valve

Precision needle valves feature an integrated stem with PTFE seat; fine pitch stem threads ensure precise adjustment.

£152.64 - £157.00

Specifications

Material Polypropylene, PTFE, Viton
Product Line Hayward NV40025T PP Needle Valve
Material (Seal) FPM O-ring
Maximum Pressure 150 psi
Max. Operating Temperature (English) 250°F
View More Specs

Products 2
Product Code Brand Outlet Port Price Quantity & Availability  
Product Code Brand Outlet Port Price Quantity & Availability  
11855342
View Documents Promotion Details
Cole-Parmer™
03245-68
3/8 in. NPT(F) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11865342
View Documents Promotion Details
Cole-Parmer™
03245-70
1/2 in. NPT(F)
£157.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-10-2024
to see stock.
Add to basket
 
Description

Description

  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
  • Valve body includes flanges for panel mounting
  • Connections are standard pipe thread
Specifications

Specifications

Polypropylene, PTFE, Viton
FPM O-ring
250°F
Hayward NV40025T PP Needle Valve
150 psi
Videos
SDS
Documents

Documents

Product Certifications