missing translation for 'onlineSavingsMsg'
Learn More

New Pig Haz-Mat Pillows

Stop the spread of acids and bases.

Brand:  New Pig PIL302

Code : Z4

Additional Details : Weight : 2.79000kg

 View more versions of this product

Product Code. 12459277

  • £113.48 / Pack of 20
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste

Specifications

Specifications

Pink
8.9 L
Hazmat Absorbent Pillow
2 x PIG™ hazmat pillows (PIL306)
8 x 8 x 1 in. (20.3 x 20.3 x 2.54 cm)
20.3 cm
20.3 cm
2.54 cm
3.4 kg
30 oz.
Polypropylene
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
8 L x 8 W x 1 in. H (20.3 x 20.3 x 2.54 cm)
8 in.
8 in.
1 in.
7.5 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers