missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

The Hazardous Substances Rack provides storage of water pollutants with a steel sump on each storage level.

Brand:  asecos™ 27829

 View more versions of this product

Product Code. 16481405

  • £797.00 / Each
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • 4 painted sumps made of steel as storage levels
Specifications

Specifications

5.3 gal.
Steel
41.73 in.
41.73 in.
78.74 in.
17.20 in.
180.78 lb.
20 L
Drum Rack
1060.00 mm
1060.00 mm
2000.00 mm
437.00 mm
82 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers