Learn More
New Pig™ HazMat Spill Kits
Brand:  New Pig™ 64CRSC
Encompass_Preferred
 View more versions of this product
Product Code. 12903595
  • £119.79 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features absorbents proven for rapid response
  • Absorbents are specially treated for unsurpassed performance with concentrated corrosives,such as 98% sulfuric acid and 30% sodium hydroxide
  • Socks stop spreading spills; PIG HazMat Pads absorb quickly
  • Prepacked container provides easy access to contents
  • Temporary disposal bags with ties help make cleanup easier
  • Tamperproof seals help prevent pilfering of spill response supplies
Specifications
Specifications
13.62 L
Spill Kit
1 External dia. 3 in., 46 in. L PIG™ HazMat Chemical Absorbent Sock, 1 External dia. 3 in., 10 ft. L PIG™ HazMat Chemical Absorbent Sock , 4 L x W: 20 x 15 in. PIG™ HazMat Mat Pad, 6 L x W: 6 x 2 in. Tamperproof Seal Label, 2 W x H: 18 x 30 in. Polyethylene Disposal Bags
Polyethylene, Polypropylene
Acids, Bases, Unknowns Fluid Absorption