Learn More
New Pig™ HazMat Spill Kits
Brand:  New Pig™ KIT320
Encompass_Preferred
 View more versions of this product
Product Code. 12913585
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Feature PIG absorbents proven for rapid response
  • Socks stop spreading spills; PIG HazMat Pads absorb quickly
  • Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bags help make cleanup easier
  • Bright yellow bag and large, color-coded label make the kit easy to locate
  • Strap for easy carrying and hanging
  • Compact and easy to store
Specifications
Specifications
9 gal.
Vinyl-laminated Nylon, Polypropylene
Acids, Bases, Unknowns
11.9 lb.
34.068 L
Spill Response Bag
4 external dia. 3 in., 46 in. L PIG&trade, HazMat Chemical Absorbent Sock, 28 L x W: 20 x 15 in. PIG&trade, HazMat Mat Pad, 5 W x H: 18 x 30 in. Polyethylene Disposal Bags, 1 Instructions
5.4 kg