Learn More
New Pig™ HazMat Spill Kits
Brand:  New Pig™ KIT220
Encompass
 View more versions of this product
Product Code. 12963615
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Feature PIG absorbents proven for rapid response
  • Socks stop spreading spills; PIG HazMat Pads absorb quickly
  • Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bags help make cleanup easier
  • Bright yellow bag and large, color-coded label make the kit easy to locate
  • Strap for easy carrying and hanging
  • Compact and easy to store
Specifications
Specifications
34.068 L
Oils, Coolants, Solvents, Water