missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Floating Foam Tube Racks
GREENER_CHOICE

Float tubes in water baths, tubs or beakers. Fisherbrand™ Floating Foam Tube Racks are constructed of flexible, high-density polyethylene foam for a variety of lab applications. Brightly colored, reusable racks include detachable carrying handles to easily lift samples out of baths.

Brand:  Fisherbrand™ HS2165B

17.04 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 11714736

  • £27.30 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Alert:

Not recommended for cleaning in autoclave.


Description

Description

  • Brightly colored foam racks are ideal for floating tubes in water baths, tubs or beakers
  • Closed cell foam resists water absorption so racks can be washed and reused
  • Include detachable carrying handles to lift samples easily out of baths
Specifications

Specifications

Not Autoclavable
Tube Rack
8 x 15 mL Tubes
High-density Polyethylene Foam
Rectangular
13 mm
Blue
For floating tubes in water baths, tubs or beakers
Detachable carrying handles
5 Pk.
17.2 x 5.7 x 6.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.