missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ HDPE General Purpose Funnels
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
Brand: Kartell™ 0017300
24.91 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
- Suitable for a wide range of liquids
- Upstand on rim reduces potential overflow
- Outer ribbing eliminates back pressure
- Hanging eye for storage
Specifications
130mm | |
20mm | |
Suitable for a wide range of liquids |
450 mL | |
HDPE | |
43mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction