missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ HDPE Industrial Tank
Kartell™ HDPE Industrial Tank is useful in everyday laboratory practice and features a comfortable handle with a secure seal-cap following DIN recommendations.
£237.00 - £285.00
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
15706978
|
Kartell™
0178100 |
5 L | GL51 |
£237.00
Pack of 40 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
10089922
|
Kartell™
0178200 |
10 L | GL51 |
£285.00
Pack of 24 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
10335443
|
Kartell™
0178000 |
2 L | GL45 | N/A | ||||||
Description
- Features a standardized industrial packaging design
- Useful in everyday laboratory practice
- Made of High Density PE food grade
- Includes a comfortable handle
- Features a secure seal-cap following DIN recommendations
- It's conform to UN regulation on transportation of dangerous and toxic liquids and is suitable for road (ADR-RID), sea (IMO-IMDG) and air (IATA-ICAO) transport
Specifications
Seal-cap | |
HDPE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kartell™ HDPE Industrial Tank