Learn More
Masterflex™ HDPE Male Pipe Adapter Barbed Fitting
For quick and easy connection of fluid paths in the laboratory without the use of tools.
Brand: Masterflex™ 30621-64
This item is not returnable.
View return policy
Description
- Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
- Available in a variety of configurations to meet your needs
- Adapts from a NPT male thread to a hose barb
- Sterilization through ethylyene oxide only
- Provided in packs of 10
Specifications
Barbed Fitting | |
3/8 NPT(M) Thread x 1/2 in. tubing I.D. | |
Ethylene Oxide | |
Straight |
HDPE | |
10 Pack | |
3/8 NPT(M) |