Learn More
Masterflex™ HDPE Male Pipe Adapter Barbed Fitting
For quick and easy connection of fluid paths in the laboratory without the use of tools.
Brand:  Masterflex™ 30621-08
 View more versions of this product
Product Code. 11799998
  • £34.90 / Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
  • Available in a variety of configurations to meet your needs
  • Adapts from a NPT male thread to a hose barb
  • Sterilization through ethylyene oxide only
  • Provided in packs of 10
Specifications
Specifications
Barbed Fitting
3/4 NPT(M) Thread x 5/8 in. tubing I.D.
Ethylene Oxide
Straight
HDPE
10 Pack
3/4 NPT(M)