missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Male Pipe Adapter Polyethylene Fittings
Combine these simple, economical fittings with flexible tubing for use in low-pressure applications (generally less than 30 psi). Cole-Parmer™ Male Pipe Adapter Polyethylene Fittings are designed for quick and easy assembly without tools.
Brand: Masterflex™ 30621-31
This item is not returnable.
View return policy
Description
- Available in a variety of materials and configurations
- Adapts from a NPT male thread to a hose barb
Specifications
Male Threaded Adapter | |
Fits 1/8 x 1/4 in. I.D. tubing | |
10/Pk. | |
Straight |
HDPE | |
-50°C | |
NPTM 0.125 in. | |
85°C |