Learn More
Labconco Heated Product Chamber
Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates. The 10 in. diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing. Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature.
Brand: Labconco 780801190
Additional Details : Weight : 30.00000kg
Beschreibung
- Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates.
- The 10 in. diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing.
- Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature from the Lyo-Works display.
- Each Chamber includes a 12 in. diameter clear acrylic cylinder, vacuum release valve, cap for vacuum port and 8 in. diameter opening to collector on bottom of chamber. Will fit 10.12 in. or 14.0 in. diameter condensers.
- Includes three shelves and three temperature probes.
- For use with any FreeZone Freeze Dryer manufactured after July 2017.
Spezifikation
Heated Product Chamber | |
-20°C to +60°C | |
Without Valves | |
230 V |
5 | |
Acrylic | |
All except North America |