missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco Heated Product Chamber
Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates. The 10 in. diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing. Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature.
£8159.24 - £13678.24
Specifications
Type | Heated Product Chamber |
---|---|
Temperature Range (Metric) | -20°C to +60°C |
Material | Acrylic |
Plug Type | All except North America |
Voltage | 230 V |
Product Code | Brand | No. of Shelves | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Shelves | Includes | Price | Quantity & Availability | ||||
17910190
|
Labconco
780801090 |
3 | Without Valves |
£8159.24
1 unit |
Estimated Shipment: 25-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17930180
|
Labconco
780801190 |
5 | Without Valves |
£11235.40
1 unit |
Estimated Shipment: 25-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17930190
|
Labconco
780802090 |
3 | Valves |
£10552.72
1 unit |
Estimated Shipment: 25-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17970180
|
Labconco
780802190 |
5 | Valves |
£13678.24
1 unit |
Estimated Shipment: 04-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates.
- The 10 in. diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing.
- Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature from the Lyo-Works display.
- Each Chamber includes a 12 in. diameter clear acrylic cylinder, vacuum release valve, cap for vacuum port and 8 in. diameter opening to collector on bottom of chamber. Will fit 10.12 in. or 14.0 in. diameter condensers.
- Includes three shelves and three temperature probes.
- For use with any FreeZone Freeze Dryer manufactured after July 2017.
Specifications
Heated Product Chamber | |
Acrylic | |
230 V |
-20°C to +60°C | |
All except North America |