missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HECW1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24695-1-AP-20UL
This item is not returnable.
View return policy
Description
HECW1 Polyclonal antibody apecifically detetects HECW1 antigen in Human, Mouse Samples. It is validated for Immunohistochemistry, ELISA.Specifications
HECW1 | |
Polyclonal | |
Unconjugated | |
BC151227 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry, ELISA | |
500 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
HECW1 | |
HECW1 fusion protein Ag20264 | |
20 μL | |
23072 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction