missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HECW1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | HECW1 |
---|---|
Concentration | 500 μg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17689606
|
Proteintech
24695-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17679606
|
Proteintech
24695-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HECW1 Polyclonal antibody apecifically detetects HECW1 antigen in Human, Mouse Samples. It is validated for Immunohistochemistry, ELISA.Specifications
HECW1 | |
Immunohistochemistry, ELISA | |
Unconjugated | |
Rabbit | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
23072 | |
HECW1 fusion protein Ag20264 | |
Primary | |
Store at -20°C. Stable for one year after shipment. |
500 μg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse | |
BC151227 | |
HECW1 | |
IgG | |
Antigen affinity purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title