Learn More
Hirschmann™ Hematocrit Tubes
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Hematocrit Tubes are manufactured from disposable capillary glass.
Brand: Hirschmann™ 9100160
This item is not returnable.
View return policy
Description
- Not heparinized
- Color-coded blue
- Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Hematocrit Tube | |
0.9/1.0 mm | |
75 ±0.5 mm |
60 μL | |
For centrifuging blood samples, not heparinized | |
DIN EN ISO 12772, CE, IVD 98/79 EC |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Hirschmann™ Hematocrit Tubes > Capacity: 60μL
Spot an opportunity for improvement?Share a Content Correction