Learn More
Hirschmann™ Hematocrit Tubes
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Hematocrit Tubes are manufactured from disposable capillary glass.
Brand:  Hirschmann™ 9100175
 View more versions of this product
Product Code. 30090389
  • £123.00 / Pack of 5000
Explore more special offers
Description
Description
  • Not heparinized
  • Color-coded blue
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Specifications
Hematocrit Tube
1.07/1.23 mm
75 ±0.5 mm
75 μL
For centrifuging blood samples, not heparinized
DIN EN ISO 12772, CE, IVD 98/79 EC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Hematocrit Tubes > Capacity: 75μL