missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hematoxylin, Fisher BioReagents
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Description
Hematoxylin is used as a stain in microscopy.

Specifications
Specifications
Melting Point | 100°C |
Bio. Stain Comm. Certification | Pass Test |
Merck Index | 15, 4672 |
Formula Weight | 302.28 |
Packaging | Glass Bottle |
Color Index No. | 75290 |
Color | Beige |
Quantity | 25 g |
Physical Form | Powder |
Chemical Name or Material | Hematoxylin |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Safety and Handling

Product Identifier
- Hematoxylin
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hematoxylin, Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction