Learn More
Hemoglobin epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12361-1-AP-20UL
 View more versions of this product
Product Code. 16810044
  • £135.00 / 20µL
Explore more special offers
Description
Description
This antibody can be used to label and isolate fetal cells from maternal blood and may be useful in prenatal diagnosis.
The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon - G-gamma - A-gamma - delta - beta-3′
Specifications
Specifications
Hemoglobin epsilon
Polyclonal
Unconjugated
HBE1
Epsilon globin, HBE, HBE1, Hemoglobin epsilon chain, Hemoglobin subunit epsilon, hemoglobin, epsilon 1
Rabbit
Antigen Affinity Chromatography
RUO
110337495, 293267, 3046
-20°C
Liquid
Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P02100
Hbe1
Hemoglobin epsilon Fusion Protein Ag3026
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hemoglobin epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated