missing translation for 'onlineSavingsMsg'
Learn More
Heptane, CHROMASOLV™, for HPLC, ≥96%, Honeywell Riedel-de Haen Product Code.: 15605890

Heptane, CHROMASOLV™, for HPLC, ≥96%, Honeywell Riedel-de Haen

Product Code. 15605890
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15605890

Brand: Honeywell-Chromasolv 5925791L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™, for HPLC,≥96%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -91°C (literature)
Absorbance max. 0.40 at 210 nm, max. 1.0 197 nm, max. 0.01 at 400 nm, max. 0.01 280 nm, max. 0.10 at 225 nm
CAS 142-82-5
CAS Max % 100.0000%
Density 0.684 g/mL at 25 °C(lit.)
Boiling Point 98.2°C to 98.4°C
Quantity 1 L
Flash Point −4°C
Packaging Glass Bottle
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
MDL Number MFCD00009544
UN Number UN1206
Beilstein 1730763
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
PubChem CID 8900
ChEBI CHEBI:43098
Formula Weight 100.2
Percent Purity ≥96%
Grade HPLC
Chemical Name or Material Heptane, CHROMASOLV™, for HPLC, ≥96%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heptane, CHROMASOLV™, for HPLC, ≥96%, Honeywell Riedel-de Haen >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.