Learn More
Heptane, ≈99% N-Heptane, for HPLC
Description
C7H16, CAS Number-142-82-5, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, heptane, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900, CCCCCCC, 3.5, 48mbar at 20 deg.C, 0.4 mPaS at 20 deg.C

Specifications
Specifications
Packaging | Amber glass bottle |
Quantity | 1 L |
Vapor Pressure | 48mbar at 20°C |
Viscosity | 0.4 mPaS at 20°C |
Vapor Density | 3.5 |
Physical Form | Liquid |
Color | Colorless |
Melting Point | -91°C |
Boiling Point | 98°C |
Formula Weight | 100.2g/mol |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527693 | Certificate of Analysis | 20/02/2025 | H/0106/30RSS,H/0106/PB17,H/0106/25RSS,H/0106/17X,H/0106/15,H/0106/17Z,H/0106/10RSS,H/0106/15X,H/0106/17 |
2527346 | Certificate of Analysis | 19/02/2025 | H/0106/30RSS,H/0106/PB17,H/0106/25RSS,H/0106/17X,H/0106/15,H/0106/17Z,H/0106/10RSS,H/0106/15X,H/0106/17 |
2527159 | Certificate of Analysis | 18/02/2025 | H/0106/30RSS,H/0106/PB17,H/0106/25RSS,H/0106/17X,H/0106/15,H/0106/17Z,H/0106/10RSS,H/0106/15X,H/0106/17 |
Safety and Handling




- Heptane
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H304-May be fatal if swallowed and enters airways.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
18Hours
20Minutes
28Seconds
33 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
49Hours
20Minutes
28Seconds
33New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.