missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, ≈99% N-Heptane, for HPLC
C7H16, CAS Number-142-82-5, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, heptane, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900, CCCCCCC, 3.5, 48mbar at 20 deg.C, 0.4 mPaS at 20 deg.C
£68.75 - £163.00
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10664912
|
Fisher Chemical
H/0106/15 |
1 L | Amber glass bottle |
£68.75
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10757704
|
Fisher Chemical
H/0106/PB17 |
2.5 L | Plastic Coated Glass bottle |
£163.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C7H16, CAS Number-142-82-5, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, heptane, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900, CCCCCCC, 3.5, 48mbar at 20 deg.C, 0.4 mPaS at 20 deg.C
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
CHEBI:43098 | |
CCCCCCC |
C7H16 | |
MFCD00009544 | |
8900 | |
heptane |
Specifications
142-82-5 | |
C7H16 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2g/mol | |
0.4 mPaS at 20°C | |
-91°C | |
Liquid |
Colorless | |
MFCD00009544 | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
48mbar at 20°C | |
3.5 | |
98°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Heptane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H304-May be fatal if swallowed and enters airways.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.