Learn More
Heptanoyl chloride, 99%, Thermo Scientific Chemicals
£30.90 - £340.00
Chemical Identifiers
CAS | 2528-61-2 |
---|---|
Molecular Formula | C7H13ClO |
Molecular Weight (g/mol) | 148.63 |
MDL Number | MFCD00000763 |
InChI Key | UCVODTZQZHMTPN-UHFFFAOYSA-N |
Synonym | enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride |
PubChem CID | 17313 |
IUPAC Name | heptanoyl chloride |
SMILES | CCCCCCC(Cl)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10669014
|
Thermo Scientific Acros
164260250 |
25 g | Glass bottle |
£30.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10502191
|
Thermo Scientific Acros
164261000 |
100 g | Glass bottle |
£73.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10761051
|
Thermo Scientific Acros
164265000 |
500 g | Glass bottle |
£340.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2528-61-2 | |
148.63 | |
UCVODTZQZHMTPN-UHFFFAOYSA-N | |
17313 | |
CCCCCCC(Cl)=O |
C7H13ClO | |
MFCD00000763 | |
enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride | |
heptanoyl chloride |
Specifications
2528-61-2 | |
Colorless to Yellow | |
173°C | |
Authentic | |
C7H13ClO | |
1.4295 to 1.4315 | |
02, 340 | |
enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride | |
UCVODTZQZHMTPN-UHFFFAOYSA-N | |
heptanoyl chloride | |
17313 | |
99% | |
Heptanoyl chloride, 99% |
-84°C | |
0.9600g/mL | |
68°C | |
98.5% min. (GC) | |
CH3(CH2)5COCl | |
MFCD00000763 | |
0.96 | |
Solubility in water: hydrolysis. Other solubilities: slightly soluble in carbontetrachloride,soluble in ethylether,ligroin | |
CCCCCCC(Cl)=O | |
148.63 | |
148.63 | |
Liquid |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-775-3
missing translation for 'tsca' : TSCA
RUO – Research Use Only