Learn More
Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinets

Herasafe 2030i BASIC, unconfigured BSC to EN12469, 230V, 50Hz. Herasafe 2030i CONFIGURED to include Cross Beam Smart UV. Additional safety features for ultimate protection and graphic user interface for ease of operation.
£14578.58 - £20935.53
Specifications
Certifications/Compliance | EN 12469 |
---|---|
Chamber Material | Stainless Steel (316) |
Product Type | Biosafety Cabinet |
Model | Herasafe 2030i |
Sash Opening (Metric) | 200 mm |
Product Code | Brand | Description | Dimensions (L x W x H) Exterior | Outlets | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Dimensions (L x W x H) Exterior | Outlets | Price | Quantity & Availability | |||
15943120
![]() |
Thermo Scientific™
51032324 |
Herasafe 2030i, 0.9 m, with 200 mm Sash Opening, No options. | 800 x 1000 x 1536 | 2 Rear Wall |
£14578.58
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15983120
![]() |
Thermo Scientific™
51032328 |
Herasafe 2030i, 0.9 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. | 800 x 1000 x 1536 | 2 Rear Wall |
£16019.49
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15993120
![]() |
Thermo Scientific™
51032329 |
Herasafe 2030i, 1.2 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. | 800 x 1300 x 1536 | 2 Rear Wall |
£16612.80
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17800325
![]() |
Thermo Scientific™
51032330 |
Herasafe 2030i, 1.5 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. | 800 x 1600 x 1536 | 4 Rear Wall |
£20426.97
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15913130
![]() |
Thermo Scientific™
51032331 |
Herasafe 2030i, 1.8 m, with 200 mm Sash Opening, Cross Beam UV in Side Walls. | 800 x 1900 x 1536 | 4 Rear Wall |
£20935.53
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinets (BSC) are designed to maximize sample protection and user safety with exceptional containment measures, while at the same time, delivering excellent user comfort and convenience. Herasafe 2030i biosafety cabinets incorporate innovative features, such as SmartFlow Plus, DAVe Plus and Smart UV technologies for excellent protection. Alarms, alerts, system self-checks and performance status are provided on a full-color touchscreen. With enabled connectivity (available in select countries) the remote app provides all data you need.
Breakthrough innovations and unprecedented features make the Herasafe 2030i BSC our most technologically advanced and user-friendly Class II, Type A2 cabinet available.
Smart protection:
- Thermo Scientific™ SmartFlow™ Plus Technology - Autonomous safety features maintain proper air flow and monitor critical conditions in real time without user interaction.
- Self-adjusting germicidal UV light intensity - Our unique cross beam UV system has exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use.
- 316L stainless steel interior - robust and resistant to scratches and chemicals
Simplified operation and ergonomics:
- Touchscreen interface
- SmartClean Plus window design – Easily access the interior of the BSC with the motorized front sash which automatically stops at the set working height. The sash can be further opened electronically or the whole front panel hinges open for thorough cleaning access.
- Bright work area with glass side walls and LED lights
- Ergonomic Arm Rests (option)
- SmartPort convenience - Easily pass tubing and wiring through sidewall to accommodate many applications needs.
Information management:
- Touchscreen and connectivity (available in select countries) – Built-in WiFi system is ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools and more. Digital Airflow Verification (DAVe) alerts, system self-checks and performance status are provided on the full-color touchscreen.
Energy efficiency:
- LED interior illumination
- Dual DC motors
- Standby mode
Certifications:
- For North America, cabinets are NSF-49 certified (4ft and 6ft models)
- For Europe, the cabinets are certified according to EN12469 (all 4 sizes). The 1.2 m model has been certified to EN 12469 with the NF mark by LNE as well.
Available sizes:
- North America 4ft and 6ft models, other countries: 0.9 m, 1.2 m, 1.5 m and 1.8 m
For cell and gene therapy applications, we offer the Herasafe 2030i Biological Safety Cabinet CTS (Cell Therapy Systems) Series which consists of the product, a comprehensive documentation package including certifications and protocols, as well as compliance services.
Previous models: Herasafe KS
Note: Please order stand separately and indicate any additional requirements/ options you need when you Request a quote
Specifications
EN 12469 | |
Biosafety Cabinet | |
200 mm | |
0.99995 | |
Energy efficient | |
2 x Each Side plus Smart Ports | |
50/60 Hz | |
230 V |
Stainless Steel (316) | |
Herasafe 2030i | |
7 in. GUI | |
H14 HEPA | |
LED | |
7.8 in. | |
800 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.