Learn More
Thermo Scientific™ Heratherm™ Refrigerated Incubators

Offers precise temperature control, increased energy efficiency, and ease of usability. Equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results.
Brand: Thermo Scientific™ 51031562
5973.29 GBP valid until 2025-12-31
Use promo code "25042" to get your promotional price.
View more versions of this product
Description
Achieve untapped potential in incubation with the temperature precision you need for applications that demand it. With a temperature range of +5 to +70°C, Thermo Scientific™ Heratherm™ Refrigerated Incubators are equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results. With no hazardous refrigerants, these units use less energy than the conventional product, making it the ideal choice for energy-conscious labs.
- Temperature range 5- 70°C
- Available in 2 sizes: benchtopmodel 178L / 6.3 cuft and a floormodel with casters 381L / 13.5 cu.ft.
- Great temperature uniformity and stability to keep the samples in a safe environment -> uniformity as low as +/-0.3°C (20°C), stability as low as +/-0.1°C
- Energy consumption as low as 80Wh/h (bench top) / 125Wh/h(floor) at 20°C - Note that comparable compressor cooled units consume up to 6 times this amount*.
- CFC free / HFC free polyurethane insulation foam for great temperature performance and condensate prevention
- Heated door with intelligent energy control to ensure no condensation – even at low temperatures
- Sophisticated timer function with weekly, daily or real time mode
- Intuitive programmability function for defined temperature ramping, soak/cool and dwell: store up to 10 programs with 10 easy steps
- Adjustable over temperature shut-off for protocols that require setting of over temperature for protection – (class 2 according to norm DIN12880)
- Safe viewing of samples through internal glass door with limited impact on temperature
- Safe containment with automatic over and under temperature alarm
- Standard access port for data monitoring (large diameter of 42mm / 1.65 in.)
- Flexible shelf system for optimal loading, easy to remove for cleaning, with non-tip protection
- Stainless steel interior (1.4301 / ASTM 304) and rounded corners for easy cleaning
Services
2 years

Specifications
6.3 cu. ft. | |
Benchtop Model | |
Stainless Steel (304/1.4301) | |
Coated Steel | |
N. America: Nema 5-15 plug w/ 120V 60Hz config.; All others: CEE7/7 with 230V 50Hz config. | |
Automated Incubator | |
Solid outer door & internal glass door | |
Base Model | |
IMP180 | |
1.6 in. (42mm) | |
0.1°C | |
543 x 464 x 708 mm (21.4 x 18.3 x 27.9 in.) | |
36.2 in. (adjustable feet: 1.4 in.- not included) | |
214 lb. | |
181 lb. | |
100/230 V 50/60 Hz, 1.9-4.5 A, 450 w |
CE, CSA | |
178 L | |
17.28 x 19.88 in. (439 x 505 mm) | |
2/9 | |
5°C to 70°C | |
Large Vacuum Fluorescent | |
Energy efficient | |
CFC free/HFC free polyurethane insulation foam | |
1 | |
Peltier technology: no refrigerant | |
≤0.2°C at 20°C; ≤0.5°C at 37°C | |
738 x 640 x 920 mm (29.1 x 25.2 x 36.2 in.) | |
920 mm (Adjustable feet: 35 mm - not included) | |
97 kg | |
82 kg | |
100/230 V |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.