Learn More
Hexacarbonylmolybdenum, 98%, Thermo Scientific Chemicals
£31.50 - £659.00
Chemical Identifiers
CAS | 13939-06-5 |
---|---|
Molecular Formula | C6MoO6 |
Molecular Weight (g/mol) | 264.01 |
MDL Number | MFCD00003466 |
InChI Key | KMKBZNSIJQWHJA-UHFFFAOYSA-N |
Synonym | molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5gShow More |
PubChem CID | 2724068 |
ChEBI | CHEBI:30508 |
IUPAC Name | carbon monoxide;molybdenum |
SMILES | [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15421037
|
Thermo Scientific Alfa Aesar
013057.88 |
2.5 g | N/A | N/A | N/A | |||||
11352138
|
Thermo Scientific Alfa Aesar
013057.14 |
25 g | N/A | N/A | N/A | |||||
11362138
|
Thermo Scientific Alfa Aesar
013057.22 |
100 g | N/A | N/A | N/A | |||||
Description
Hexacarbonylmolybdenum is widely utilized in electron beam-induced deposition technique providing an appropriate source of molybdenum atoms. It finds an application as important reagent in organometallic synthesis because of replacement of carbonyl ligands by other donor ligands. Its derivative is act as a catalyst in organic synthesis, which is used in alkyne metathesis and the Pauson-Khand reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13939-06-5 | |
264.01 | |
KMKBZNSIJQWHJA-UHFFFAOYSA-N | |
2724068 | |
carbon monoxide;molybdenum |
C6MoO6 | |
MFCD00003466 | |
molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5gShow More | |
CHEBI:30508 | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo] |
Specifications
∽150°C (decomposition) | |
Crystalline | |
C6MoO6 | |
MFCD00003466 | |
molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g | |
KMKBZNSIJQWHJA-UHFFFAOYSA-N | |
carbon monoxide;molybdenum | |
2724068 | |
264 | |
1.96 |
13939-06-5 | |
98% | |
Mo(CO)6 | |
UN3466 | |
Slightly soluble in tetrahydrofuran,diglyme and acetonitrile. | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo] | |
264.01 | |
CHEBI:30508 | |
98% | |
Hexacarbonylmolybdenum, Up to 1% Cr |
Safety and Handling
GHS H Statement
H300-H330
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 237-713-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.