Learn More
Hexacarbonylmolybdenum, 98%, Thermo Scientific Chemicals
Description
Hexacarbonylmolybdenum is widely utilized in electron beam-induced deposition technique providing an appropriate source of molybdenum atoms. It finds an application as important reagent in organometallic synthesis because of replacement of carbonyl ligands by other donor ligands. Its derivative is act as a catalyst in organic synthesis, which is used in alkyne metathesis and the Pauson-Khand reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∽150°C (decomposition) |
Physical Form | Crystalline |
Quantity | 2.5 g |
Assay Percent Range | 98% |
Linear Formula | Mo(CO)6 |
UN Number | UN3466 |
Solubility Information | Slightly soluble in tetrahydrofuran,diglyme and acetonitrile. |
Formula Weight | 264 |
Percent Purity | 98% |
Density | 1.96 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P17L041 | Certificate of Analysis | 19/03/2025 | 013057.88,013057.22,013057.14 |
T01K053 | Certificate of Analysis | 05/08/2024 | 013057.22,013057.88,013057.14 |
P08K018 | Certificate of Analysis | 14/03/2024 | 013057.22,013057.88,013057.14 |
X02G011 | Certificate of Analysis | 19/09/2022 | 013057.22,013057.88,013057.14 |
Y17H012 | Certificate of Analysis | 12/04/2022 | 013057.22,013057.88,013057.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.