missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilazane, Electronic grade, 99+%, Thermo Scientific Chemicals

£24.62 - £184.92

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
MDL Number MFCD00008259
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11309215
SDS View Documents
Thermo Scientific Alfa Aesar
L16519.AC
25 mL
£24.62
25mL
Please to purchase this item. Need a web account? Register with us today!
11319215
SDS View Documents
Thermo Scientific Alfa Aesar
L16519.AE
100 mL
£56.88
100mL
Please to purchase this item. Need a web account? Register with us today!
11329215
SDS View Documents
Thermo Scientific Alfa Aesar
L16519.AP
500 mL
£184.92
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

Solubility
Miscible with methanol, ethyl ether, chloroform and benzene. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.  
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H331-H314-H318-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H311-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 213-668-5

missing translation for 'rtecsNumber' : JM9230000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexamethyldisilazane, Electronic grade, 99+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.