missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene™ High Efficiency Particulate Air (HEPA) Filter
High Efficiency Particulate Air (HEPA) Filters are available in various configurations for the containment of particles.
Brand: Syngene™ SYN-ACFHEPA18W
Additional Details : Weight : 3.62870kg
Description
- The filter has a 99.997% efficiency rating for collecting 0.3 micron particles.
Specifications
99.997% | |
3 in. | |
18 in. | |
Wood Frame | |
46 cm |
HEPA | |
12 in. | |
8 cm | |
30.5 cm |