Learn More
Greiner Bio-One High Grade Polypropylene Pipet Tips
Manufactured from high-grade polypropylene to give the optimum performance and fit. Greainer BioOne™ High Grade Polypropylene Pipet Tips are ideally suited for Gilson™ Pipetman models.
Brand:  Greiner Bio-One 745290
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11789542
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • High breaking strength, dimensionally stable, heat-resistant up to approx. +140°C; cold-resistant down to -190°C
  • Autoclavable
  • Free of heavy metals
  • Conforms to the ISO 9001 standard
  • Tested for conformance with the international standard DIN ISO 8655
  • Liquid end is bevelled so less liquid remains on pipet tip, contact surface of pipetted liquids is reduced and pipetting accuracy increases
  • Ideally suited for Gilson™ Pipetman P20, P100, P200, F5, F10, F25, F50, F200, U200
Specifications
Specifications
1000 to 5000 μL
Autoclavable
Bulk
Polypropylene
Natural
ISO 9001, DIN ISO 8655
Socorex™ Pipette, Gilson™ Pipetman™ P5000