missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiLoad™ 16/600 Superdex™ 200 pg
Use for separating MAbs from dimers and contaminants. Cytiva HiLoad™ 16/600 Superdex™ 200 pg is ideal for high-resolution preparative gel filtration chromatography.
Brand: Cytiva 28-9893-35
2079.55 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Includes:
- Dismantling tool
- Support screen
- Net ring (10μm)
- O-ring
- Stop plugs
- Instructions
Description
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose. Dextran component determined the separation properties of these media. Steep selectivity curves give gppd resolving power for peptides and proteins in the molecular weight range Mr 10 000 600 000 (Superdex 200 pg).- Used for separating MAbs from dimers and contaminants
- Short run times with high recovery
- Medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions
- Separations can be scaled up to production levels
- 0.06 in. fittings to connect to AKTA design systems
- Theoretical plates: >13000m-1
- Flow rate: <1.6mL/min.
- Pressure max. (over the packed bed during operation): 3 bar [0.3MPa] (42psi)
- Media
- Average particle size: 34μm
- Matrix: composite of cross-linked agarose and dextran
- pH Stability working range: 3 to 12
- pH Stability cleaning range: 2 to 14
- Fractionation [Mr] globular proteins: 1 × 104-6 × 105
- Fractionation [Mr] dextrans: 1 × 103-1 × 105
- Shelf life from manufacture date: 5 years
- Chemical stability: all commonly used buffers, 1M NaOH, 0.1M HCl, 1M acetic acid, 8M urea, 6M guanidine HCl, 1% SDS, 70% ethanol, 30% iso-propanol, 30% acetonitrile
- Column
- Column I.D.: 0.62 in. (16mm)
- Inlet I.D.: 0.031 in. (0.8mm)
- Outlet I.D.: 0.031 in. (0.8mm)
- Chemical stability: resistant to aqueous solutions and solvents used in liquid chromatography. Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol
- Column hardware made from borosilicate glass, polyamide, fuororubber, Ethylene Propylene Diene (EPDM), ETFE, Superpolyoxymethylene
- Column tube made from borosilicate glass
- Bed support made from Polypropylene (PP) (reinforced glass fiber), Polyamide (PA)
- Porosity: 10μm
Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids
Order Info
HiLoad columns are prepacked XK columns; Bed volume: 120mL; Bed dimensions: 0.62 x 23.62 in. (16 x 600mm); Sample volume: 5mL
Specifications
<5 mL | |
4°C to 30°C, 20% Ethanol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiLoad 16/600 Superdex 200 pg
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol