Learn More
Cytiva HiLoad™ Superdex™ 200 PG XK Column
Prepacked XK columns designed for high-resolution preparative gel filtration chromatography.
£4256.00 - £9210.00
Product Code | Brand | Bed Volume | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Volume | Price | Quantity & Availability | |||||
11380502
|
Cytiva
90-1002-73 |
480mL |
£5817.00
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
11389991
|
Cytiva
90-1001-37 |
180mL |
£4256.00
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
11369991
|
Cytiva
90-1000-40 |
1000mL |
£9210.00
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
Description
HiLoad™ Superdex™ 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice for separating MAbs from dimers and contaminants.
- High resolution with short run times and good recovery.
- High capacity at high flow rates.
- Separations can be scaled up to production levels.
- Prepacked for convenience and reproducibility.
- 1/16” fittings for convenient connections to ÄKTA design systems.
Superdex™ prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these resins are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex™ 30 pg), Mr 3 000 to 70 000 (Superdex™ 75 pg), and Mr 10 000 600 000 (Superdex™ 200 pg). The resin combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.
Specifications
4 to 30°C, 20% Ethanol |