missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ HIC FF 16/10 Columns

Produce fast, preparative separation of proteins and other biomolecules. Cytiva Life Sciences™ HiPrep™ HIC FF 16/10 Columns are high capacity with excellent flow properties, and reliable, reproducible separations.

Brand:  Cytiva 28-9365-45

Code : GE

Additional Details : Weight : 2.10920kg


 View more versions of this product

Product Code. 10608225

  • £592.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

The 20mL HIC columns provide fast, preparative separation of proteins and other biomolecules. They are ready-to-use with four media choices: Phenyl Sepharose 6 Fast Flow (high sub); Phenyl Sepharose 6 Fast Flow (low sub); Butyl Sepharose 4 Fast Flow; and Octyl Sepharose 4 Fast Flow.

Features:

  • Offer high capacity, excellent flow properties and reliable, reproducible separations
  • Compatible with simple pump-based configurations or chromatography systems (e.g., AKTA)

Specifications (H2O at room temp.)

  • Dimensions: 1.6 x 10cm
  • Recommended flowrate: 2-10mL/min. (60-300cm/h)
  • Maximum flowrate: 10mL/min. (300cm/hr.)
  • Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
  • Column hardware pressure limit: 5bar (73psi, 0.5MPa)
  • Storage: 20% ethanol
  • Storage temperature: 4° to 30°C

Specifications

Specifications

1M NaOH, 3M Ammonium Sulphate, 30% Isopropanol, 70% Ethanol, 10% Ethylene Glycol, 0.5% SDS, 6M Guanidine Hydrochloride, 8M Urea
20mL
16mm
3.93 in.
>300cm/hr.
Phenyl
3 to 13
6% Cross-linked Agarose
Ion Exchange Column
HiPrep Phenyl Sepharose 4 FF (high sub) column
0.62 in.
1 x 20mL
100mm
Polypropylene
5 bar
45 to 165μm
4° to 30°C
Provide fast, preparative separation of Proteins and other biomolecules by hydrophobic interaction chromato graphy (HIC), initial and intermediate purification, removing bulk impurities
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiPrep Phenyl FF (high sub) 16/10

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers