missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ IEX FF 16/10 Columns

Provide fast, reliable and reproducible preparative ion exchange separations

Brand:  Cytiva 28-9365-42

Code : GE

Additional Details : Weight : 0.10200kg


 View more versions of this product

Product Code. 10617855

  • £567.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

DEAE and CM Sepharose™ 6 Fast Flow are prepacked in 20mL HiPrep columns. Ideal for rapid enrichments during initial capture of proteins from starting material.

Features:

  • Combine good flowrates and high binding capacity
  • Simple-to-operate
  • Compatible with single-pump configurations and other chromatography systems (e.g., AKTA™)

Specifications (H2O at room temp.)

  • Dimensions: 1.6 x 10cm
  • Recommended flowrate: 2-10mL/min. (60-300cm/h)
  • Maximum flowrate: 10mL/min. (300cm/hr.)
  • Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
  • Column hardware pressure limit: 5bar (73psi, 0.5MPa)
  • Storage: 20% ethanol (DEAE,CM), 0.2M sodium acetate in 20% ethanol (SP)
  • Storage temperature: 4° to 30°C

Specifications

Specifications

1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
20mL
16mm
3.93 in.
>300cm/hr.
Carboxymethyl
6 to 10
6% Cross-linked Agarose
Ion Exchange Column
HiPrep CM Sepharose FF column
0.62 in.
1 x 20mL
100mm
Polypropylene
1.5 bar
45 to 165μm
4° to 30°C
Preparative separations of Proteins and other biomolecules using ion exchange (IEX) chromato graphy
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiPrep CM FF 16/10

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers