missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ IMAC FF 16/10 Column

For optimizing purification of histidine-tagged proteins at laboratory scale

Brand:  Cytiva 28-9365-52

Code : GE

Additional Details : Weight : 0.11000kg


Product Code. 10237654

  • £424.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

IMAC Sepharose™ 6 Fast Flow is prepacked in 20mL HiPrep columns for greater scale up. The medium consists of 90μm beads of highly crosslinked agarose, to which a chelating group has been covalently coupled. This group may be charged with suitable metal ions, specific for your target protein.The bead size enables high flow properties; low metal ion leakage retains activity of the purified protein.

Specifications:
IMAC Sepharose 6 Fast Flow

  • Matrix: highly crosslinked agarose, 6%
  • Average particle size: 90μm
  • Binding capacity (protein/ion dependent): approx. 40mg histidine-tagged protein/mL medium when charged with Ni2+
  • Compatibility: stable in all commonly used buffers
  • Chemical stability (uncharged medium) one week, 40°C: 0.01 M HCl, 0.1 M NaOH; 12 hr.: 1M NaOH, 70% acetic acid; 1 hr. tested 2% SDS; 30 min. tested 30% 2-propanol
  • Stability (pH, w/ uncharged medium w/ metal ion): 3 to 12 (long-term), 2 to 14 (short-term)
  • Storage/Temperature: 20% ethanol; 4° to 30°C

HiPrep Prepacked Columns

  • Volume: 20mL
  • Dimensions: 1.6 x 10cm
  • Max. pressure during operation: 1.5bar (0.15MPa, 22psi)
  • Hardware pressure limit: 5bar (0.5MPa, 73psi)
  • Recommend flowrate (H2O at room temp.): 2 to 10mL/min.
  • Specifications

    Specifications

    0.01M HCl, 1M NaOH, 70% Acetic Acid, 2% SDS, 30% 2-propanol
    20mL
    16mm
    3.93 in.
    <10mL/min.
    1.5 bar
    45 to 165μm
    4° to 30°C
    IMAC and subsequent purification of Polyhistidine tagged Proteins
    HiTrap IMAC Sepharose 6 FF column
    0.62 in.
    1 x 20mL
    100mm
    Polypropylene
    3 to 12
    6% Cross-linked Agarose
    Ion Exchange Column
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • HiPrep IMAC FF 16/10

    Signal Word
    • Warning

    Hazard Category
    • Flammable liquid Category 3

    Hazard Statement
    • H226-Flammable liquid and vapour.

    Precautionary Statement
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

    Supplemental information
    • MIXTURE LIST-Contains : ethanol

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers