Learn More
Cytiva HiPrep™ Sephacryl™ HR Prepacked Gel Filtration Columns
For reproducible protein purification, separating polysaccharides, macromolecules with extended structures and small plasmids
Brand: Cytiva 28-9356-04
Code : GX
Additional Details : CAS Number : 64-17-5 Weight : 0.43700kg
Description
Aids in preparative separation of biomolecules from up to 5mL (for 16mm column I.D.) and 13mL (for 26mm column I.D.).
- Provides maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability
- Prepacked columns for use with AKTAdesign™ or other pumped chromatography systems
- Available in two different prepacked column sizes - 16 x 600mm (120mL) and 26 x 60mm (320mL)
- Maximum pressure (over packed bed during operation) is 1.5 bar (22psi)
Media
- Average particle size: 50μm
- Matrix: Allyl dextran and N, N'-methylenebisacrylamide
- pH Stability working range: 3 to 11
- pH Stability cleaning range: 2 to 13
- Chemical stability: all commonly used buffers, 0.2 M NaOH, 0.1 M HCl, 1 M acetic acid, 8 M urea, 6 M guanidine HCl, 1% SDS, 20% ethanol, 30% propanol, 30% acetonitrile (tested at 40°C for one week), 0.5 M NaOH (only for cleaning-in-place)
- Pressure/Flow specification: >125cm/hr., 100kPa, XK 50/30, bed height 15cm
- Material (bed support): Polymethyl-Methacrylate
HiPrep 16/60 Sephacryl S-400 HR
- Media:
- Fractionation [Mr] globular proteins: 2 x 104-8 x 106
- Exclusion limit [Mr] (globular proteins): ≈9 x 106 (9000kDa)
- Fractionation range [Mr] DNA exclusion limit: 271bp
- Fractionation [Mr] Dextrans: 1 x 104-2 x 106
- Media:
- Exclusion limit for Dextrans: 100 x 106
- Fractionation range [Mr] DNA exclusion limit: 1078bp
- Fractionation [Mr] Dextrans: 4 x 104-2 x 107
Specifications
Each | |
25 to 75 μm | |
120 mL | |
1.6cm | |
23.62 in. | |
Polypropylene | |
3 to 11 | |
Polysaccharides, macromolecules with extended structures and even small plasmids |
< 0.5 mL/min. | |
Low to medium pressure AKTA™ systems or pumped chromatography systems | |
Sephacryl HR Pre-packed Gel Filtration Column | |
Reproducible Protein purification, separating Polysaccharides, macromolecules with extended structures and small plasmids | |
60cm | |
1.5 bar [0.15 MPa] (22 psi) | |
Allyl Dextran and N,N'-methylenebisacrylamide | |
4 to 30°C, 20% Ethanol |
Safety and Handling
- HiPrep 16/60 Sephacryl S-400 HR
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol