missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Sephacryl™ S-100 HR Columns

Maximum recovery and minimum nonspecific adsorption

Brand:  Cytiva 17-1194-01

Code : A0

Additional Details : Weight : 0.70500kg


 View more versions of this product

Product Code. 10369334

  • £975.00 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Recommended linear flowrate 5 to 15cm/hr.

Designed for separating peptides and small proteins

Specifications

Specifications

Each
25 to 75 μm
320 mL
2.6cm
23.62 in.
Polypropylene
3 to 11
Peptides and small proteins
< 1.3 mL/min.
Low to medium pressure AKTA™ systems
Sephacryl HR Column
Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix
60cm
1.5 bar [0.15 MPa] (22 psi)
Allyl Dextran and N, N'-methylenebisacrylamide
4°C to 30°C, 20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiPrep 26/60 Sephacryl S-100 HR

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers