missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiPrep™ Sephacryl™ S-300 HR Columns
Maximum recovery and minimum nonspecific adsorption
£673.00 - £975.00
Specifications
Quantity | Each |
---|---|
Particle Size | 25 to 75 μm |
For Use With (Equipment) | Low to medium pressure AKTA™ systems |
Product Type | Sephacryl HR Column |
For Use With (Application) | Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix |
Product Code | Brand | Bed Volume | Diameter (Metric) Inner | Flow Rate | Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Volume | Diameter (Metric) Inner | Flow Rate | Format | Price | Quantity & Availability | ||
10616695
|
Cytiva
17-1167-01 |
120mL | 1.6cm | <0.5 mL/min. | Bed Dimensions: 16x60 |
£673.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
10702817
|
Cytiva
17-1196-01 |
320mL | 2.6cm | <1.3 mL/min. | Bed Dimensions: 26x60 |
£975.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
Description
- Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
- Recommended linear flowrate 5 to 15cm/hr.
- Designed for purifying antibodies, serum proteins and midsize proteins
Specifications
Each | |
Low to medium pressure AKTA™ systems | |
Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix | |
60cm | |
22psi | |
Antibodies, serum proteins and mid-size proteins |
25 to 75 μm | |
Sephacryl HR Column | |
23.62 in. | |
Polypropylene | |
3 to 11 | |
4° to 30°C |