Learn More
Cytiva HiPrep™ Sephacryl™ S-400 HR Columns
Maximum recovery and minimum nonspecific adsorption
Brand: Cytiva 28-9356-05
Code : GX
Additional Details : CAS Number : 64-17-5 Weight : 0.66500kg
Description
- Long-term chemical and physical stability from hydrophilic, rigid allyl dextran/bisacrylamide matrix
- Recommended linear flowrate 5 to 15cm/hr.
Specifications
Each | |
25 to 75 μm | |
320 mL | |
2.6cm | |
23.62 in. | |
Polypropylene | |
3 to 11 | |
Polysaccharides, macromolecules with extended structures, small plasmids |
< 1.3 mL/min. | |
Low to medium pressure AKTA™ systems | |
Sephacryl HR Column | |
Separation of biomolecules with maximum recovery and minimum nonspecific adsorption with long term chemical and physical stability resulting from the hydrophilic, rigid Allyl Dextran/Bisacrylamide matrix | |
60cm | |
1.5 bar [0.15 MPa] (22 psi) | |
Allyl Dextran and N, N'-methylenebisacrylamide | |
4 to 30°C, 20% Ethanol |
Safety and Handling
- HiPrep 26/60 Sephacryl S-400 HR
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol