Learn More
Cytiva His Mag Sepharose™ Ni Magnetic Beads
Designed for efficient, small-scale purification/screening of histidine-tagged proteins from different sources Cytiva Lifescience™ His Mag Sepharose™ Ni Magnetic Beads enable high-capacity small-scale purification.
Brand: Cytiva 29104065
Description
Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel. For higher throughput, larger numbers of samples can easily be screened in parallel on a robotic device.
- High-capacity small-scale purification of histidine-tagged proteins from different sources
- High purity and yield
- Easy parallel screening of histidine-tagged proteins with high repeatability
- Scalability dash (en) Simple capture of histidine-tagged proteins from small or large sample volumes (low microliter to milliliter scale)
Order Info
1mL of medium slurry is sufficient for five purification runs according to the recommended protocol
Specifications
50 mg Histidine-tagged protein/mL medium | |
100 mL |
Safety and Handling
- His Mag Sepharose Ni, 100 ml
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, nickel
- UNKNOWN ACUTE-66.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.