Learn More
Cytiva His-tag Purification Kit
For use with ÄKTA Start, and the purification of Histidine-tagged proteins
Brand: Cytiva 29-0588-03
Code : NEW
Additional Details : Weight : 0.11000kg
Description
- User-friendly prepacked columns for shorter start-up time
- Column packages adapted for specific purification needs
- High performance media for reliable purification results
Specifications
HisTrap™ HP (1 x 1 mL), HiTrap™ TALON™ Crude (1 x 1 mL), and HiTrap™ Desalting (1 x 5 mL) | |
Purification of Histidine-Tagged Proteins | |
Protein Purification System |
ÄKTA Start | |
His-tagged |
Safety and Handling
- Tagged-Package His, (29051021/29048565/29048684)
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.