missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScale™ Columns Packed with Superdex™ Increase or Superose™ Increase Resins

Similar to the new-generation Superdex™ Increase and Superose™ Increase Size Exclusion Chromatography (SEC) Columns, but for use with purifying larger volumes of up to 8.5 mL

Brand:  Cytiva 29321906

Additional Details : CAS Number : 64-17-5 Weight : 2.70000kg


 View more versions of this product

Product Code. 15938872

  • £9590.00 / Each
Estimated Shipment: 09-09-2024
to see stock.
Add to basket

Description

Description

  • High-resolution SEC columns
  • Small particle size and narrow particle size distribution of the packed resin ensures highly pure protein
Specifications

Specifications

Size Exclusion Chromatography (SEC)
1 Ea.
8.5 mL
Superdex 200 Increase
2 MPa
Store at 4°C to 30°C, 20% Ethanol
26 x 400 mm
Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers