Learn More
Cytiva HiScreen™ Capto™ Q ImpRes and Capto™ SP ImpRes
High-resolution intermediate purification and polishing based on the well-established Capto platform with traditional ligands
Brand: Cytiva 17-5470-02
373.35 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
- Delivers pressure-flow properties with wide resolution
- High-throughput purifications easy to optimize and scale up
- High flow agarose based matrix
- Runs at high flow rates as well as high bed heights thus increasing flexibility in process design
- Developed for large-scale purification of biopharmaceutical proteins
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
- Available in a wide range of different bulk pack sizes and pre-packed formats for process development
Capto Q ImpRes: Strong anion (quaternary amine group) exchange BioProcess™ medium
Capto SP ImpRes: Strong cation (sulfonate group) exchange BioProcess medium
Specifications
1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol | |
39.3 in. | |
100 mL | |
200mm | |
Quaternary Amine | |
2 to 12 | |
High-flow Agarose | |
High-throughput intermediate purification and polishing steps of a wide range of biomolecules |
Capto Q ImpRes medium | |
1000mm | |
7.87 in. | |
220 cm/hr. | |
<3 bar | |
36 to 44μm | |
4° to 30°C | |
Capto Q Chromatography Media |
Safety and Handling
- capto Q impres
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.