Learn More
Cytiva™ HiScreen™ Column with MabSelect™ VL Protein L Resin
These HiScreen™ columns contain Protein L resin for affinity capture of antibody Fabs, bispecifics, and other variants containing a kappa light chain.
Brand: Cytiva™ 17542015
Additional Details : CAS Number : 64-17-5 Weight : 0.08550kg
Description
As the diversity of antibody variants increases, so does the need for chromatography resins that can capture and purify these variants. As an alternative to protein A, protein L resin can be used to capture antibody Fabs, bispecific antibodies, and other molecules containing kappa variants of the variable light (VL) chain. Protein L binds human kappa light chain subtypes 1, 3, and 4 and mouse kappa light chain subtype 1.
Redesigned protein L ligand MabSelect™ VL protein L resin showed a two-fold increase in binding capacity compared to Cytiva™ Capto™ L chromatography resin. This higher capacity provides productivity advantages in research and biomanufacturing.
- A redesigned protein L ligand captures antibody Fabs, bispecifics, and other variants containing a kappa light chain with high binding capacity and improved alkaline stability
- High dynamic binding capacity for efficient capture of target molecules
- Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents
- Provides good resolution for product-related impurities in the capture of bispecific antibodies
- Convenient for method optimization and parameter screening enabled by the 10 cm bed height for efficient chromatography process development
Specifications
70 mg Human IgG/mL Resin at 6 min. Residence Time, 60 mg Human IgG/mL Resin at 4 min. Residence Time | |
0.3 in. | |
<300 cm/hr. | |
60 μm | |
1 Column | |
Antibody | |
Chromatography Column | |
Affinity |
4.7 mL | |
7.7 mm | |
3 bar | |
HiScreen™ MabSelect™ | |
Antibody Affinity Chromatography | |
Pre-packed Column | |
100 mm |