missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ IMAC Sepharose™ 6 Prepacked Fast Flow Columns
Designed for method optimization and parameter screening
Brand: Cytiva 28-9505-17
This item is not returnable.
View return policy
Description
- Based on 6% highly cross-linked agarose with an average bead size of 90μm
- Processed flow properties with high binding and loading capacities
- Bed height of 10cm is designed to allow method optimization and parameter screening
- Optimal bead size for scale up
- Charge with the metal of choice for purification of histidine-tagged proteins
- Reproducible results, scalable to BioProcess™ columns packed with the same chromatography media using the same linear fluid velocity
- Bed volume: 4.7mL
- Bed dimension: 7.7 × 100mm
- Matrix: 6% cross-linked agarose beads
- Average particle size: 90μm
- Max. flow velocity: <600cm/hr
- Material (Column hardware and column tube): polypropylene
- Column I.D.: 7.7mm
Specifications
0.01M HCl, 1M NaOH, 70% Acetic Acid, 2% SDS, 30% 2-propanol | |
4.7mL | |
7.7mm | |
3.93 in. | |
<600cm/hr. | |
1.5 bar | |
45 to 165μm | |
4° to 30°C | |
Optimization and parameter screening |
HiScreen IMAC IMAC Sepharose 6 FF column | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
3 to 12 | |
6% Cross-linked Agarose | |
Ion Exchange Column |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiScreen IMAC FF
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction