missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Q Sepharose™ and SP Sepharose™ Fast Flow IEX Columns
Designed for method optimization and parameter screening
Brand: Cytiva 28-9505-10
This item is not returnable.
View return policy
Description
- Ready-to-use column for convenient process development optimization and parameter screening
- Highly cross-linked agarose matrix with average bead size of 90μm
- Bed height of 10cm designed to allow method optimization and parameter screening
- 20cm bed height is acheived when it is connected in series
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
- Bed dimension: 7.7 × 100mm
- Matrix: 6% cross-linked agarose beads
- Average particle size: 90μm
- Max. flow velocity: 450cm/hr.
- Material (column hardware and column tube): polypropylene
- Column I.D.: 7.7mm
HiScreen Q FF
- Media:
- pH stability working range: 2 to 12
- pH stability cleaning: 2 to 14
- Ion exchanger type: strong anion exchanger
- Media:
- pH stability working range: 3 to 14
- pH stability cleaning: 4 to 13
- Ion exchanger type: strong cation exchanger
Specifications
1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol | |
4.7mL | |
7.7mm | |
3.93 in. | |
<450 cm/hr. | |
Quaternary Amine | |
2 to 12 | |
6% Cross-linked Agarose | |
Ion Exchange Column |
HiScreen Q Sepharose FF Column | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
1.5 bar | |
45 to 165μm | |
4° to 30°C | |
Optimization and parameter screening, ion exchange chromato graphy |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction