missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Q Sepharose™ and SP Sepharose™ High Performance IEX Columns
Designed for method optimization and parameter screening
£122.00
Specifications
Buffer | 8M Urea, 6M Guanidine HCl, 70% Ethanol, 1M NaOH, 1M Acetic Acid, 30% Isopropanol, 30% Aceto nitrile, 2% SDS |
---|---|
Bed Volume | 4.7mL |
Diameter (English) Inner | 0.3 in. |
Diameter (Metric) Inner | 7.7mm |
Quantity | 1 x 4.7mL |
Product Code | Brand | Product Type | Ligand Type | pH Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Ligand Type | pH Range | Price | Quantity & Availability | |||
10397509
|
Cytiva
28-9505-11 |
HiScreen Q Sepharose HP column | Quaternary Amine | 2 to 12 |
£122.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10870911
|
Cytiva
28-9505-15 |
HiScreen SP Sepharose HP column | Sulphopropyl | 4 to 13 |
£122.00
Each |
Estimated Shipment: 20-09-2024
Log in to see stock. |
|||
Description
- Ready-to-use columns for convenient process development optimization and parameter screening
- Highly cross-linked agarose matrix with an average bead size of 34μm
- Bed height of 10cm designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
- Bed dimension: 7.7 × 100mm
- Matrix: 6% cross-linked agarose beads
- Average particle size: 34μm
- Max. flow velocity: <150cm/hr.
- Material (column hardware and column tube): polypropylene
- Column I.D.: 7.7mm
- Flow velocity: <150cm/hr.
HiScreen Q HP
- Ionic capacity: 0.14 to 0.20mmol Cl-/mL
- Ligand: quaternary amine
- Ion exchanger type: strong anion exchanger
- Ionic capacity: 0.15 to 0.20mmol Cl-/mL
- Ligand: sulphopropyl
- Ion exchanger type: strong cation exchanger
Specifications
8M Urea, 6M Guanidine HCl, 70% Ethanol, 1M NaOH, 1M Acetic Acid, 30% Isopropanol, 30% Aceto nitrile, 2% SDS | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
45 to 165μm | |
4° to 30°C | |
Optimization and parameter screening, ion exchange chromato graphy |
4.7mL | |
7.7mm | |
3.93 in. | |
<150cm/hr. | |
3 bar | |
6% Cross-linked Agarose | |
Ion Exchange Column |