Learn More
Thermo Scientific™ HisPur™ Cobalt Resin
Gently purify His-tagged recombinant fusion proteins using cobalt (Co2+) immobilized metal chelate affinity (IMAC) beads, spin columns and plates.
£113.00 - £3115.00
Specifications
Content And Storage | Store at 4°C |
---|---|
Form | Liquid Suspension |
Product Line | HisPur™ |
Protein Form | Recombinant |
Target | His-Tagged Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
10751375
|
Thermo Scientific™
89964 |
10 mL | N/A | N/A | N/A | |||||
10270054
|
Thermo Scientific™
89965 |
100 mL |
£746.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10344300
|
Thermo Scientific™
89966 |
500 mL |
£3115.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.
Additional Information and Online Ordering
HisPur Cobalt Resins, Columns and Kits use a tetradentate chelating resin charged with divalent cobalt (Co2+)to enable specific, fast and gentle purification of histidine-tagged fusion proteins from a variety of sample types. HisPur Cobalt Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high throughput needs. With these options, high-quality His-tag protein purification is possible at nearly any scale. HisPur Cobalt Resin is superior to traditional nickel-based immobilized metal affinity chromatography (IMAC) systems. Compared to Ni-IDA and tetradentate Ni2+chelate resins, HisPur Resin binds histidine-tagged proteins with higher specificity (less off-target binding) and releases them with gentler conditions (lower concentrations of imidazole). Use HisPur Columns and Cartridges to obtain high-purity His-tagged proteins with no metal contamination.Highlights:
High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions
Low metal leaching – no metal contamination in eluted histidine-tagged protein sample
Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives
Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates
Cost effective – resin can be reused several times or discarded after single use
Superior – performs better than other commercially available IMAC Resins
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
- Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin
Includes:
Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer
Specifications
Store at 4°C | |
HisPur™ | |
His-Tagged Protein | |
Metal Chelate, Divalent Cobalt | |
Affinity, Agarose Resin |
Liquid Suspension | |
Recombinant | |
Resin | |
Bottle |
Safety and Handling
- HisPur Cobalt Resin
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H350-May cause cancer.
- H360F-May damage fertility.
- H411-Toxic to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P273-Avoid release to the environment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Cobalt chloride(CoCl2), hexahydrate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.