missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Ni-NTA Magnetic Beads

Product Code. 13484289 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
10mL
2mL
This item is not returnable. View return policy

Product Code. 13484289

Brand: Thermo Scientific™ 88832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform affinity purification of His-tagged fusion proteins in manual or automated formats with the high capacity nickel-IMAC Thermo Scientific™ HisPur Ni-NTA Magnetic Beads.

Thermo Scientific HisPur Ni-NTA Magnetic Beads are high-capacity nickel-IMAC beads for affinity purification of His-tagged fusion proteins in manual or automated formats.

Features of HisPur Ni-NTA Magnetic Beads:

  • High capacity—equivalent or higher binding capacity than Ni-NTA magnetic beads from other suppliers
  • Low nonspecific binding—the bead surface is pre-blocked and the protocol provides optimized buffers for purification
  • Fast—protocol is completed in 1 hour
  • Scalable—process microliter to milliliter sample volumes
  • Versatile—purify proteins using native or denaturing conditions
  • Reagent compatible—can be used with common cell lysis reagents and a variety of buffer additives
  • Multiple formats—protein coupling to the beads and downstream applications can be performed both manually and on an automated platform (e.g., Thermo Scientific KingFisher Instruments)

The blocked magnetic bead surface is derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) beads provide high binding capacity with very low background. The HisPur Ni-NTA Magnetic Beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher Instruments for high-throughput needs.

HisPur Ni-NTA Magnetic Beads are used for small scale affinity purification as well as high-throughput screening of recombinant His-tagged proteins. The polyhistidine tag is the most popular affinity tag and typically consists of six consecutive histidine residues (6xHis). These tagged proteins are overexpressed in a number of different systems, most commonly in bacteria, and purified from cell lysates such as those prepared using B-PER Bacterial Protein Extraction Reagents. Purification of His-tagged proteins is achieved using a NTA chelate charged with nickel that coordinates with the histidine side chains. The NTA chelate contains four metal-binding sites which allow for low metal ion leaching and high binding capacity. The protocol for the HisPur Ni-NTA Magnetic Beads has been optimized to allow for high purity of the isolated His-tagged protein. Performance is equivalent to or better than Ni-NTA magnetic beads from other suppliers.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 mL
Concentration Slurry: 12.5mg/mL, 1.25% solids
Content And Storage Store at 4°C
Particle Size 1 μm
Product Line HisPur
Quantity 10 mL
Protein Form Recombinant
Target His
Type Magnetic Affinity Bead
Ligand Type Nickel-NTA
compliance-icons compliance-icons
Product Identifier
  • HisPur Ni-NTA Magnetic Beads
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, sulphuric acid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.